missing translation for 'onlineSavingsMsg'
Learn More

D(-)-Isoascorbic Acid (Erythorbic Acid), TRC

Product Code. 17928224
Change view
Click to view available options
Quantity:
10 g
100 g
250 g
Unit Size:
100g
10g
250g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17928224 10 g 10g
30012812 250 g 250g
30004715 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17928224 Supplier Toronto Research Chemicals Supplier No. TRCE64989010G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 89-65-6
Molecular Formula C6 H8 O6
Molecular Weight (g/mol) 176.12
Synonym D-erythro-Hex-2-enonic acid, γ-lactone, D-erythro-Hexonic acid, 3-keto-, γ-lactone (6CI), Erythorbic acid (7CI), Araboascorbic acid, Araboascorbic acid, D-, D-(-)-Isoascorbic acid, D-Araboascorbic acid, D-Erythorbic acid, D-Isoascorbic acid, D-arabino-Ascorbic acid, E 315, Erycorbin, Glucosaccharonic acid, Isoascorbic acid, Isovitamin C, Mercate 5, NSC 8117, Neo-Cebicure, Saccharosonic acid, D-Isoascorbic Acid, (5R)-5-[(1R)-1,2-Dihydroxyethyl]-3,4-dihydroxyfuran-2(5H)-one, Ascorbic Acid Imp. F (EP)
IUPAC Name (2R)-2-[(1R)-1,2-dihydroxyethyl]-3,4-dihydroxy-2H-furan-5-one
SMILES OC[C@@H](O)[C@H]1OC(=O)C(=C1O)O

Specifications

Quantity 10 g
InChI Formula InChI=1S/C6H8O6/c7-1-2(8)5-3(9)4(10)6(11)12-5/h2,5,7-10H,1H2/t2-,5-/m1/s1
Formula Weight 176.0321
Chemical Name or Material Isoascorbic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.