missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D(-)-Isoascorbic acid, 98%

Product Code. 10267283 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Unit Size:
2.5kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10267283 2.5 kg 2.5kg
10491651 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10267283 Supplier Thermo Scientific Chemicals Supplier No. 227270025
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 89-65-6 | C6H7NaO6 | 198.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 89-65-6
Molecular Formula C6H7NaO6
Molecular Weight (g/mol) 198.11
MDL Number MFCD00005378
InChI Key IFVCRSPJFHGFCG-HXPAKLQESA-N
Synonym erythorbic acid, isoascorbic acid, d-araboascorbic acid, d-isoascorbic acid, araboascorbic acid, d-erythorbic acid, isovitamin c, neo-cebicure, saccharosonic acid, mercate 5
PubChem CID 54675810
ChEBI CHEBI:51438
IUPAC Name (2R)-2-[(1R)-1,2-dihydroxyethyl]-3,4-dihydroxy-2H-furan-5-one
SMILES [Na+].OC[C@H](O)C1OC(=O)[C-](O)C1=O

Specifications

Melting Point 167.0°C to 172.0°C
Color White to Yellow
Loss on Drying 0.4% max.
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (Iodimetry)
Packaging Plastic bottle
Quantity 2.5 kg
Merck Index 15, 5171
Specific Rotation Condition −17.25°(25°C c=10,H2O)
Specific Rotation − 17.25
Solubility Information Solubility in water: 100g/l. Other solubilities: soluble in alcohol,pyridine,moderately soluble in acetone,slightly soluble in glycerol
Formula Weight 176.13
Percent Purity 98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material D(-)-Isoascorbic acid, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.