missing translation for 'onlineSavingsMsg'
Learn More

D-α-Hydroxyglutaric acid disodium, MedChemExpress

Product Code. 16459636
Encompass_Preferred
Click to view available options
Quantity:
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
This item is not returnable. View return policy

Product Code. 16459636

Brand: Medchem Express HY100542

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress D-α-Hydroxyglutaric acid disodium (Disodium (R)-2-hydroxyglutarate) is the principal metabolite accumulating in neurometabolic disease D-2-hydroxyglutaric aciduria.

D-α-Hydroxyglutaric acid disodium is a weak competitive antagonist of α-ketoglutarate (α-KG) and inhibits multiple α-KG-dependent dioxygenases with a Ki of 10.87 mM. D-α-Hydroxyglutaric acid disodium increases reactive oxygen species (ROS) production. D-α-Hydroxyglutaric acid disodium binds and inhibits ATP synthase and inhibits mTOR signaling.

Specifications

Chemical Name or Material D-α-Hydroxyglutaric acid disodium
Physical Form Powder
CAS 103404-90-6
Color Off-White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C5H6Na2O5
For Use With (Application) Cancer-Kinase/protease
Synonym Disodium (R)-2-hydroxyglutarate
Solubility Information H2O : 75 mg/mL (390.46 mM; Need ultrasonic)
SMILES O=C(O[Na])[C@H](O)CCC(O[Na])=O
Molecular Weight (g/mol) 192.08
Formula Weight 192.08
Percent Purity 95.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.