missing translation for 'onlineSavingsMsg'
Learn More

D-Homophenylalanine, 98%, Thermo Scientific Chemicals

Catalog Number 11426580
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
82795-51-5
C10H13NO2
179.22
MFCD00063091
JTTHKOPSMAVJFE-SECBINFHSA-N
d-homophenylalanine, h-d-hophe-oh, 2r-2-amino-4-phenylbutanoic acid, r-2-amino-4-phenylbutanoic acid, d-homophenyl-ala, h-d-homophe-oh, r-homophenylalanine, r-2-amino-4-phenylbutyric acid, unii-5847zw3pep, d-homophenyl alanine
6950302
(2R)-2-amino-4-phenylbutanoic acid
N[C@H](CCC1=CC=CC=C1)C(O)=O
This item is not returnable. View return policy
82795-51-5
C10H13NO2
179.22
MFCD00063091
JTTHKOPSMAVJFE-SECBINFHSA-N
d-homophenylalanine, h-d-hophe-oh, 2r-2-amino-4-phenylbutanoic acid, r-2-amino-4-phenylbutanoic acid, d-homophenyl-ala, h-d-homophe-oh, r-homophenylalanine, r-2-amino-4-phenylbutyric acid, unii-5847zw3pep, d-homophenyl alanine
6950302
(2R)-2-amino-4-phenylbutanoic acid
N[C@H](CCC1=CC=CC=C1)C(O)=O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point >300°C
Beilstein 4675530
Formula Weight 179.22
Percent Purity 98%
Quantity 1 g
Chemical Name or Material D-Homophenylalanine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'