missing translation for 'onlineSavingsMsg'
Learn More

D-Homocystine, TRC

Product Code. 30402170
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30402170 1 g 1g
30395661 10 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30402170 Supplier Toronto Research Chemicals Supplier No. TRCH5911901G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6027-15-2
Molecular Formula C8H16N2O4S2
Molecular Weight (g/mol) 268.35
Synonym (2R,2'R)-4,4'-Dithiobis[2-aminobutanoic Acid, NSC 206271
IUPAC Name (2R)-2-amino-4-[[(3R)-3-amino-3-carboxypropyl]disulfanyl]butanoic acid
SMILES N[C@H](CCSSCC[C@@H](N)C(=O)O)C(=O)O

Specifications

InChI Formula InChI=1S/C8H16N2O4S2/c9-5(7(11)12)1-3-15-16-4-2-6(10)8(13)14/h5-6H,1-4,9-10H2,(H,11,12)(H,13,14)/t5-,6-/m1/s1
Quantity 1 g
Formula Weight 268.0551
Shipping Condition Room Temperature
Chemical Name or Material D-Homocystine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.