missing translation for 'onlineSavingsMsg'
Learn More

D-Glutamine, MedChemExpress

Product Code. 16499646
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16499646 50 mg 50mg
16474386 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16499646 Supplier Medchem Express Supplier No. HY100587
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress D-Glutamine is a cell-permeable D type stereoisomer of Glutamine.

D-Glutamine is a cell-permeable D type stereoisomer of Glutamine.

Specifications

Chemical Name or Material D-Glutamine
CAS 5959-95-5
Color White
Purity Grade Notes Research
Molecular Formula C5H10N2O3
Solubility Information H2O : ≥ 32 mg/mL (218.97 mM)
SMILES N[C@H](CCC(N)=O)C(O)=O
Molecular Weight (g/mol) 146.14
Quantity 50 mg
Formula Weight 146.14
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.