missing translation for 'onlineSavingsMsg'
Learn More

D-Glutamine, 99.2%, MP Biomedicals™

Product Code. 11344249 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11344249 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11344249 Supplier MP Biomedicals Supplier No. 0210180110
 Restricted Item
Estimated Shipment: 12-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Unnatural isomer of glutamine suitable for cell analysis research applications.

Specifications

CAS 5959-95-5
Color White
Assay Percent Range 0.992
Molecular Formula C5H10N2O3
Packaging Glass Bottle
Synonym d-glutamine, h-d-gln-oh, d-2-aminoglutaramic acid, l +-glutamine, 2r-2-amino-4-carbamoylbutanoic acid, r-2,5-diamino-5-oxopentanoic acid, d-glutamic acid 5-amide, d-glutamin, unii-63hb36ca2y, d-gln
InChI Key ZDXPYRJPNDTMRX-GSVOUGTGSA-N
SMILES C(CC(=O)N)C(C(=O)O)N
IUPAC Name (2R)-2,5-diamino-5-oxopentanoic acid
Molecular Weight (g/mol) 146.146
Quantity 10 g
PubChem CID 145815
ChEBI CHEBI:17061
Formula Weight 146.1
Percent Purity ≥98%
Grade Biochemical
Physical Form Powder
Chemical Name or Material D-Glutamine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.