missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D(-)-Glutamine, 98%

Product Code. 10271592 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10271592 5 g Glass bottle 5g
10068671 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10271592 Supplier Thermo Scientific Chemicals Supplier No. 135160050
 Restricted Item
Estimated Shipment: 05-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 5959-95-5 | C5H10N2O3 | 146.146 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5959-95-5
Molecular Formula C5H10N2O3
Molecular Weight (g/mol) 146.146
MDL Number MFCD00065607
InChI Key ZDXPYRJPNDTMRX-GSVOUGTGSA-N
Synonym d-glutamine, h-d-gln-oh, d-2-aminoglutaramic acid, l +-glutamine, 2r-2-amino-4-carbamoylbutanoic acid, r-2,5-diamino-5-oxopentanoic acid, d-glutamic acid 5-amide, d-glutamin, unii-63hb36ca2y, d-gln
PubChem CID 145815
ChEBI CHEBI:17061
IUPAC Name (2R)-2,5-diamino-5-oxopentanoic acid
SMILES C(CC(=O)N)C(C(=O)O)N

Spezifikation

Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula H2NCOCH2CH2CH(NH2)COOH
Quantity 5 g
Specific Rotation Condition −32.00 (20.00°C 589nm,c=10,N HCl)
Specific Rotation -30° to -34° (20°C, 589nm) (c=10, 2N HCl)
Formula Weight 146.15
Percent Purity ≥97.5%
Physical Form Crystalline Powder
Chemical Name or Material D(-)-Glutamine, 98%
Mehr anzeigen Weniger anzeigen

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.