missing translation for 'onlineSavingsMsg'
Learn More

D-Glucuronic Acid, TRC

Product Code. 30011584
Change view
Click to view available options
Quantity:
100 mg
10 g
25 g
Unit Size:
100mg
10g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30011584 25 g 25g
30008372 10 g 10g
17999254 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30011584 Supplier Toronto Research Chemicals Supplier No. TRCG59685025G
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6556-12-3
Molecular Formula C6 H10 O7
Molecular Weight (g/mol) 194.14
Synonym D-Glucuronic Acid, Glucuronic acid, D- (8CI), D-(+)-Glucuronic acid, Glucosiduronic acid, Glucuronic acid
IUPAC Name (2S,3S,4S,5R)-2,3,4,5-tetrahydroxy-6-oxohexanoic acid
SMILES O[C@@H](C=O)[C@@H](O)[C@H](O)[C@H](O)C(=O)O

Specifications

Quantity 25 g
InChI Formula InChI=1S/C6H10O7/c7-1-2(8)3(9)4(10)5(11)6(12)13/h1-5,8-11H,(H,12,13)/t2-,3+,4-,5-/m0/s1
Formula Weight 194.0427
Percent Purity >95%
Chemical Name or Material D-Glucuronic Acid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.