missing translation for 'onlineSavingsMsg'
Learn More

D-(+)-Glucose monohydrate, BAKER ANALYZED™, J.T. Baker™, Avantor™

Product Code. 10447080 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10447080 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10447080 Supplier JT Baker Supplier No. 0113.1000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Laboratory Reagent

Specifications

CAS 14431-43-7
Melting Point 83°C
Ignition Residue max. 0.015%
Molecular Formula C6H12O6·H2O
MDL Number MFCD00149450
Quantity 1 kg
Molecular Weight (g/mol) 198.17
Insoluble Matter max. 0.005%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.