missing translation for 'onlineSavingsMsg'
Learn More

D(+)-Glucose 6-phosphate sodium salt, 98%

Product Code. 11986901 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11986901 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11986901 Supplier Thermo Scientific Acros Supplier No. 446980010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 54010-71-8 | C6H12NaO9P | 282.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 54010-71-8
Melting Point 192°C to 197°C
Assay Percent Range 98%
Packaging Glass Bottle
Molecular Formula C6H12NaO9P
Quantity 1 g
Specific Rotation + 33
Synonym sodium 2r,3r,4s,5r-2,3,4,5-tetrahydroxy-6-oxohexyl hydrogenphosphate, sodium glucose 6-phosphate, robison ester monosodium salt, d-glucose-6-phosphate monosodium salt, d-glucose, 6-dihydrogen phosphate , monosodium salt, d +-glucose 6-phosphate sodium salt, d-glucose 6-phosphoric acid hydrogen sodium salt, sodium 2r,3s,4r,5r-6-hydrogen phosphonatooxy-2,3,4,5-tetrahydroxyhexanal, sodium 2r,3r,4s,5r-2,3,4,5-tetrahydroxy-6-oxo-hexyl hydrogen phosphate
InChI Key OBHLNVXMRZXIII-BTVCFUMJSA-M
SMILES C(C(C(C(C(C=O)O)O)O)O)OP(=O)(O)[O-].[Na+]
IUPAC Name sodium;[(2R,3R,4S,5R)-2,3,4,5-tetrahydroxy-6-oxohexyl] hydrogen phosphate
Molecular Weight (g/mol) 282.12
PubChem CID 23677332
Formula Weight 282.12
Percent Purity 98%
Chemical Name or Material D(+)-Glucose 6-phosphate sodium salt
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.