missing translation for 'onlineSavingsMsg'
Learn More

D-Glucose-6-Phosphate, Disodium Salt Hydrate, MP Biomedicals™

Product Code. p-7083075 Shop All MP Biomedicals Products
Click to view available options
Quantity:
500 mg
5 g
25 g
Unit Size:
25g
500mg
5g
This item is not returnable. View return policy

Product Code. 11462990

Brand: MP Biomedicals 02100312.5

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Lies at the start of the glycolysis and pentose phosphate metabolic pathways. D-Glucose-6-Phosphate, Disodium Salt Hydrate, MP Biomedicals is the core carbohydrate substrate for the enzymatic synthesis of archaetidyl-myo-inositols produced through myo-ino

Glucose 6-phosphate (also known as Robison ester) is glucose sugar phosphorylated on carbon 6. This compound is very common in cells, as the vast majority of glucose entering a cell will become phosphorylated in this way. Because of its prominent position in cellular chemistry, glucose 6-phosphate has many possible fates within the cell.

Glucose 6-phosphate lies at the start of two major metabolic pathways: Glycolysis and Pentose phosphate. In addition to these metabolic pathways, glucose 6-phosphate may also be converted to glycogen or starch for storage. This storage is in the liver and muscles in the form of glycogen for most multicellular animals, and in intracellular starch or glycogen granules for most other organisms.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3671-99-6
Molecular Formula C6H11Na2O9P
Molecular Weight (g/mol) 304.10
MDL Number MFCD00136037
InChI Key CHIBNKGHYAQTQY-OGPXQWFKNA-L
Synonym d-glucose-6-phosphate disodium salt, sodium 2r,3r,4s,5r-2,3,4,5-tetrahydroxy-6-oxohexyl phosphate, sodium glucose 6-phosphate, disodium d-glucose 6-phosphate, glucose 6-phosphate sodium salt, glucose-6-phosphate disodium, disodium d-glucose-6-phosphate, d-glucose 6-disodium phosphate, d-glucose 6-phosphoric acid disodium salt, disodium 2r,3s,4r,5r-2,3,4,5-tetrahydroxy-6-phosphonooxy hexanal
PubChem CID 18594434
IUPAC Name disodium;[(2R,3R,4S,5R)-2,3,4,5-tetrahydroxy-6-oxohexyl] phosphate
SMILES [Na+].[Na+].O[C@H]1O[C@H](COP([O-])([O-])=O)[C@@H](O)[C@H](O)[C@H]1O

Specifications

pH 7 to 9 (1% aq. soln.)
Quantity 500 mg
Solubility Information Soluble in distilled water or dilute buffer.
Optical Rotation +30–38° (c=1, water)
Formula Weight 304.2 (anhydrous)
Percent Purity ≥98%
Physical Form Powder
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.