missing translation for 'onlineSavingsMsg'
Learn More

D-Glucose-6-phosphate disodium salt dihydrate, Thermo Scientific™

Product Code. 15865148 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
5 g
10 g
100 g
250 g
Packaging:
Plastic bottle
Unit Size:
100g
10g
250g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15865148 250 g Plastic bottle 250g
15813761 5 g Plastic bottle 5g
15845148 10 g Plastic bottle 10g
15855148 100 g Plastic bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15865148 Supplier Thermo Scientific Supplier No. J16185.30

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3671-99-6
Molecular Formula C6H11Na2O9P
Molecular Weight (g/mol) 304.10
MDL Number MFCD00136037
InChI Key CHIBNKGHYAQTQY-OGPXQWFKNA-L
Synonym d-glucose-6-phosphate disodium salt, sodium 2r,3r,4s,5r-2,3,4,5-tetrahydroxy-6-oxohexyl phosphate, sodium glucose 6-phosphate, disodium d-glucose 6-phosphate, glucose 6-phosphate sodium salt, glucose-6-phosphate disodium, disodium d-glucose-6-phosphate, d-glucose 6-disodium phosphate, d-glucose 6-phosphoric acid disodium salt, disodium 2r,3s,4r,5r-2,3,4,5-tetrahydroxy-6-phosphonooxy hexanal
PubChem CID 18594434
SMILES [Na+].[Na+].O[C@H]1O[C@H](COP([O-])([O-])=O)[C@@H](O)[C@H](O)[C@H]1O

Specifications

Physical Form Powder
Quantity 250 g
IUPAC Name disodium;[(2R,3R,4S,5R)-2,3,4,5-tetrahydroxy-6-oxohexyl] phosphate
Formula Weight 340.13
Packaging Plastic bottle
Chemical Name or Material D-Glucose-6-phosphate disodium salt dihydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.