missing translation for 'onlineSavingsMsg'
Learn More

D-(+)-Glucosamine Hydrochloride, 99%, MP Biomedicals™

Product Code. 11314219 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11314219 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11314219 Supplier MP Biomedicals Supplier No. 0210178290

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Molecular Biology
TRUSTED_SUSTAINABILITY

Specifications

CAS 66-84-2
Assay Percent Range 99%
Molecular Formula C6H13NO5.ClH
MDL Number MFCD00135831
Quantity 500 g
Synonym 2-Amino-2-deoxy-D-glucose hydrochloride, Chitosamine hydrochloride
Formula Weight 215.6
Physical Form Solid
Chemical Name or Material D-(+)-Glucosamine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.