missing translation for 'onlineSavingsMsg'
Learn More

D-Glucosamine-13C Hydrochloride, TRC

Product Code. 30398552
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30398552 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30398552 Supplier Toronto Research Chemicals Supplier No. TRCG51495225MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material D-Glucosamine-13C Hydrochloride
CAS 84247-63-2
Molecular Formula 13C C5 H13 N O5 . Cl H
Quantity 25 mg
Synonym D-Glucose-1-13C, 2-amino-2-deoxy-, hydrochloride (9CI), 2-Amino-2-deoxy-D-glucose-1-13C hydrochloride, D-Glucosamine-1-13C Hydrochloride
InChI Formula InChI=1S/C6H13NO5.ClH/c7-3-5(10)4(9)2(1-8)12-6(3)11;/h2-6,8-11H,1,7H2;1H/t2-,3-,4-,5-,6?;/m1./s1/i6+1;
SMILES Cl.N[C@H]1[13CH](O)O[C@H](CO)[C@@H](O)[C@@H]1O
IUPAC Name (3R,4R,5S,6R)-3-amino-6-(hydroxymethyl)(2^{13}C)oxane-2,4,5-triol;hydrochloride
Molecular Weight (g/mol) 216.625
Formula Weight 216.059
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.