missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-Gluconic Acid Potassium Salt, TRC
Click to view available options
Quantity:
5 g
Unit Size:
5g
Specifications
Specifications
| Chemical Name or Material | Potassium Gluconate |
| CAS | 299-27-4 |
| Molecular Formula | C6 H11 O7 . K |
| Quantity | 5 g |
| Synonym | Potassium Gluconate |
| InChI Formula | InChI=1S/C6H12O7.K/c7-1-2(8)3(9)4(10)5(11)6(12)13;/h2-5,7-11H,1H2,(H,12,13);/q;+1/p-1/t2-,3-,4+,5-;/m1./s1 |
| SMILES | [K+].OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C(=O)[O-] |
| IUPAC Name | potassium;(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate |
| Molecular Weight (g/mol) | 234.25 |
| Formula Weight | 234.0142 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction