missing translation for 'onlineSavingsMsg'
Learn More

D-Gluconic Acid (50% in Water), TRC

Product Code. 30403207
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30403207 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30403207 Supplier Toronto Research Chemicals Supplier No. TRCG4174201G
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material D-Gluconic Acid
CAS 526-95-4
Molecular Formula C6 H12 O7
Quantity 1 g
Synonym Dextronic Acid, Gluconal GA-50, Gluconic Acid, Glycogenic Acid, Glyconic Acid, Maltonic Acid, NSC 77381, Pentahydroxycaproic Acid
InChI Formula InChI=1S/C6H12O7/c7-1-2(8)3(9)4(10)5(11)6(12)13/h2-5,7-11H,1H2,(H,12,13)/t2-,3-,4+,5-/m1/s1
SMILES OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C(=O)O
IUPAC Name (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoic acid
Molecular Weight (g/mol) 196.1553
Formula Weight 196.0583
Concentration or Composition (by Analyte or Components) 0.5
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.