missing translation for 'onlineSavingsMsg'
Learn More

D-Glucaro-1,4-lactone Monohydrate, TRC

Product Code. 30392836
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30392836 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30392836 Supplier Toronto Research Chemicals Supplier No. TRCG415600250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material D-Glucaro-1,4-lactone Monohydrate
CAS 61278-30-6
Purity Grade Notes HPLC
Molecular Formula C6 H8 O7 . H2 O
Quantity 250 mg
InChI Formula InChI=1S/C6H8O7.H2O/c7-1-2(8)6(12)13-4(1)3(9)5(10)11;/h1-4,7-9H,(H,10,11);1H2/t1-,2-,3+,4+;/m1./s1
SMILES O.O[C@@H]([C@H]1OC(=O)[C@H](O)[C@H]1O)C(=O)O
IUPAC Name (2S)-2-[(2S,3R,4R)-3,4-dihydroxy-5-oxooxolan-2-yl]-2-hydroxyacetic acid;hydrate
Molecular Weight (g/mol) 210.14
Formula Weight 210.0376
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.