missing translation for 'onlineSavingsMsg'
Learn More

D-Galactose (White to Off-white Powder), Fisher BioReagents

Product Code. 11423883
Change view
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11423883 500 g Poly Bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11423883 Supplier Fisher BioReagents Supplier No. BP656500

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
D-Galactose is suitable for use in cell culture systems requiring sugar additives.
TRUSTED_SUSTAINABILITY

Specifications

Alpha Vector Galactose
CAS 59-23-4
Melting Point 167°C
Color White
Assay Percent Range ≥98 %
Packaging Poly Bottle
Molecular Formula C6H12O6
Quantity 500 g
Merck Index 15, 4365
Specific Rotation 78.2 to 82.2° (+ or -)
Formula Weight 180.08
Percent Purity ≥98%
Physical Form Solid
Chemical Name or Material D-Galactose
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.