missing translation for 'onlineSavingsMsg'
Learn More

D-Galactose, MedChemExpress

Product Code. 16472366
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mg
5 g
Unit Size:
500mg
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16472366 500 mg 500mg
16421338 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16472366 Supplier Medchem Express Supplier No. HYN0210
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress D-Galactose is a natural aldohexose and C-4 epimer of glucose.

D-Galactose is a natural aldohexose and C-4 epimer of glucose.

Specifications

Chemical Name or Material D-Galactose
CAS 59-23-4
Color White
Purity Grade Notes Research
Molecular Formula C6H12O6
Quantity 500 mg
Synonym D-(+)-Galactose
Solubility Information H2O : 62.5 mg/mL (346.91 mM; ultrasonic and warming and heat to 60°C) ∣DMSO : 50 mg/mL (277.53 mM; Need ultrasonic)
SMILES O=C[C@@H]([C@H]([C@H]([C@@H](CO)O)O)O)O
Molecular Weight (g/mol) 180.16
Formula Weight 180.16
Percent Purity 95.0%
Grade Research
Physical Form Powder
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.