missing translation for 'onlineSavingsMsg'
Learn More

D-(+)-Galactose Anhydrous, cell culture reagent, 98.5%, MP Biomedicals™

Product Code. 11455172 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
Unit Size:
100g
1kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11455172 1 kg 1kg
11435172 100 g 100g
11445172 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11455172 Supplier MP Biomedicals Supplier No. 0219467091

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 10257-28-0
Molecular Formula C6H12O6
Molecular Weight (g/mol) 180.16
InChI Key WQZGKKKJIJFFOK-UHFFFAOYNA-N
Synonym d-galactose, d-galactopyranose, galactose, galactopyranose, galactopyranoside, d-+-galactose, galactose, pure, cerebrose, gal, d-galactopyranoside
PubChem CID 6036
ChEBI CHEBI:4139

Specifications

Quantity 1 kg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.