missing translation for 'onlineSavingsMsg'
Learn More

D-Galactal Triacetate, TRC

Product Code. 30240951
Change view
Click to view available options
Quantity:
10 g
50 g
100 g
Unit Size:
100g
10g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30240951 50 g 50g
30240518 100 g 100g
30240960 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30240951 Supplier Toronto Research Chemicals Supplier No. TRCG15200050G
 Restricted Item
Estimated Shipment: 31-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4098-06-0
Molecular Formula C12 H16 O7
Molecular Weight (g/mol) 272.25
IUPAC Name [(2R,3R,4R)-3,4-diacetyloxy-3,4-dihydro-2H-pyran-2-yl]methyl acetate
SMILES CC(=O)OC[C@H]1OC=C[C@@H](OC(=O)C)[C@H]1OC(=O)C

Specifications

Quantity 50 g
InChI Formula InChI=1S/C12H16O7/c1-7(13)17-6-11-12(19-9(3)15)10(4-5-16-11)18-8(2)14/h4-5,10-12H,6H2,1-3H3/t10-,11-,12-/m1/s1
Formula Weight 272.0896
Chemical Name or Material D-Galactal, Triacetate (syrup)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.