missing translation for 'onlineSavingsMsg'
Learn More

D-Fructose, TRC

Product Code. 30011768
Change view
Click to view available options
Quantity:
10 mg
5 g
50 g
Unit Size:
10mg
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30011768 5 g 5g
17919344 10 mg 10mg
17907524 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30011768 Supplier Toronto Research Chemicals Supplier No. TRCF7925005G
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57-48-7
Molecular Formula C6H12O6
Molecular Weight (g/mol) 180.16
Synonym Fructose, (-)-d-Arabino-hex-2-ulopyranose, D(-)-Fructose
IUPAC Name (2RS,3S,4R,5R)-2-(hydroxymethyl)oxane-2,3,4,5-tetrol
SMILES O[C@]1(CO)O[C@H](CO)[C@@H](O)[C@@H]1O

Specifications

Quantity 5 g
InChI Formula InChI=1S/C6H12O6/c7-1-3-4(9)5(10)6(11,2-8)12-3/h3-5,7-11H,1-2H2/t3-,4-,5+,6-/m1/s1
Formula Weight 180.06
Percent Purity >95%
Chemical Name or Material D-(-)-Fructose
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.