missing translation for 'onlineSavingsMsg'
Learn More

β-D-(-)-Fructose, MP Biomedicals

Product Code. 11359039 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11359039 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11359039 Supplier MP Biomedicals Supplier No. 02102186.5
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

  • Fructose and other monosaccharides are used for the diagnosis of cancer as cancer cells have shown a higher level of fructose utilization as compared to glucose

Specifications

CAS 57-48-7
Melting Point 110°C to 120°C
Molecular Formula C6H12O6
Linear Formula C6H12O6
MDL Number MFCD00148910
Quantity 500 g
Synonym d---fructose, d--fructose, 3s,4r,5r-1,3,4,5,6-pentahydroxyhexan-2-one, furucton, arabino-hexulose, keto-d-fructose, d-levulose, sugar, fruit, fructose, d, krystar 300
Solubility Information Soluble in water and alcohols.
InChI Key BJHIKXHVCXFQLS-UYFOZJQFSA-N
SMILES C(C(C(C(C(=O)CO)O)O)O)O
IUPAC Name (3S,4R,5R)-1,3,4,5,6-pentahydroxyhexan-2-one
Molecular Weight (g/mol) 180.156
PubChem CID 5984
ChEBI CHEBI:48095
Formula Weight 180.2
Percent Purity ≥99%
Physical Form Powder
Chemical Name or Material β-D-(-)-FRUCTOSE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.