missing translation for 'onlineSavingsMsg'
Learn More

D-Fructose 6-Phosphate-13C6 Sodium Salt Hydrate, TRC

Product Code. 30401189
Change view
Click to view available options
Quantity:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30401189 1 mg
30389211 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30401189 Supplier Toronto Research Chemicals Supplier No. TRCF7925781MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula 13C6H13O9P • xNa • xH2O
Molecular Weight (g/mol) 266.0922991802
SMILES O[13C@H]1[13C@H](O)[13C](O)([13CH2]O)O[13C@@H]1[13CH2]OP(O)(O)=O

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C6H13O9P/c7-2-6(10)5(9)4(8)3(15-6)1-14-16(11,12)13/h3-5,7-10H,1-2H2,(H2,11,12,13)/t3-,4-,5+,6?/m1/s1/i1+1,2+1,3+1,4+1,5+1,6+1
Chemical Name or Material D-Fructose 6-Phosphate-13C6 Sodium Salt Hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.