missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-Fructose 6-Phosphate-13C6 Disodium Salt, TRC
Specifications
Specifications
| Chemical Name or Material | D-Fructose 6-Phosphate-13C6 Disodium Salt |
| Molecular Formula | 13C6 H11 Na2 O9 P |
| Quantity | 10 mg |
| Synonym | 6-(Dihydrogen phosphate) D-Fructose Sodium Salt, D-fructose 6-phosphate Disodium |
| InChI Formula | InChI=1S/C6H13O9P.2Na/c7-2-6(10)5(9)4(8)3(15-6)1-14-16(11,12)13;;/h3-5,7-10H,1-2H2,(H2,11,12,13);;/q;2*+1/p-2/t3-,4-,5+,6?;;/m1../s1/i1+1,2+1,3+1,4+1,5+1,6+1;; |
| SMILES | O[13CH2][13C]1(O)O[13C@H]([13CH2]OP(=O)(O[Na])O[Na])[13C@@H](O)[13C@@H]1O |
| IUPAC Name | [sodiooxy-[[(2R,3S,4S)-3,4,5-trihydroxy-5-(hydroxy(113C)methyl)(2,3,4,5-13C4)oxolan-2-yl](113C)methoxy]phosphoryl]oxysodium |
| Molecular Weight (g/mol) | 310.055 |
| Formula Weight | 310.014 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?