missing translation for 'onlineSavingsMsg'
Learn More

D-Fructose-5-13C, TRC

Product Code. 30398243
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30398243 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30398243 Supplier Toronto Research Chemicals Supplier No. TRCF7925055MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material D-Fructose-5-13C
CAS 635325-97-2
Molecular Formula C513CH12O6
Quantity 5 mg
Synonym Advantose FS 95-5-13C, D-(-)-Fructose-5-13C, D-(-)-Levulose-5-13C, D-Arabino-2-hexulose-5-13C, Fructose-5-13C, Fruit Sugar-5-13C, Fujifructo L 95-5-13C, Furucton-5-13C, Hi-Fructo 970-5-13C, Krystar-5-13C, Krystar 300-5-13C, Levulose-5-13C, Nevulose-5-13C, Sugar Fruit-5-13C
SMILES O[13C@@H]1[C@H](O)[C@@H](O)C(O)(CO)OC1
Molecular Weight (g/mol) 181.15
Formula Weight 181.0667
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.