missing translation for 'onlineSavingsMsg'
Learn More

Melford D-Fructose

Product Code. 30018276
Change view
Click to view available options
Quantity:
500 g
1 kg
Unit Size:
1kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30018276 1 kg 1kg
30018277 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30018276 Supplier Melford Supplier No. F215001000.0

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Product usually ships within 24 hours. Research or further manufacturing use only, not for food or drug use.

  • Substance Name: Levulose
  • CAS Number: 57-48-7
  • Molecular Formula: C6H12O6
  • Molecular Weight: 180.16

Specifikationer

CAS 57-48-7
Molecular Formula C6H12O6
Quantity 1 kg
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.