missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D-Fructose 1,6-bisphosphate trisodium salt octahydrate, 98%

Product Code. 15420537
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
Unit Size:
100g
10g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15420537 100 g 100g
15400537 10 g 10g
15410537 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15420537 Supplier Thermo Scientific Chemicals Supplier No. J66256.22
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 81028-91-3 | C6H26Na3O20P2 | 549.17 g/mol

D-Fructose-1,6-bisphosphate (FBP), a common metabolic sugar, is the precursor of glyceraldehyde 3-phosphate and dihydroxyacetone phosphate in the glycolytic pathway. It is an allosteric activator of enzymes such as pyruvate kinase and a substrate used to identify and characterize enzymes such as fructose-1,6-bisphosphate aldolase(s) and fructose-1, 6-bisphosphatase(s). FBP is studied as a neuroprotective agent in brain injury.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-Fructose-1,6-bisphosphate (FBP), a common metabolic sugar, is the precursor of glyceraldehyde 3-phosphate and dihydroxyacetone phosphate in the glycolytic pathway. It is an allosteric activator of enzymes such as pyruvate kinase and a substrate used to identify and characterize enzymes such as fructose-1,6-bisphosphate aldolase(s) and fructose-1, 6-bisphosphatase(s). FBP is studied as a neuroprotective agent in brain injury.

Solubility
Soluble in water

Notes
Moisture Sensitive, store away from water/moisture. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 81028-91-3
Molecular Formula C6H26Na3O20P2
Molecular Weight (g/mol) 549.17
MDL Number MFCD00149340,MFCD00135870,MFCD00068321
InChI Key DBTJWGHAMYSWQR-OLMHPWOBSA-J
Synonym FDP-Na3H
IUPAC Name trisodium octahydrate [(3S,4S,5R)-2,3,4-trihydroxy-5-[(phosphonatooxy)methyl]oxolan-2-yl]methyl phosphate
SMILES O.O.O.O.O.O.O.O.[Na+].[Na+].[Na+].O[C@H]1[C@H](O)C(O)(COP([O-])([O-])=O)O[C@@H]1COP([O-])([O-])=O

Specifications

Color White
Quantity 100 g
Beilstein 3837985
Sensitivity Moisture sensitive
Solubility Information Soluble in water
Formula Weight 550.18
Percent Purity 98%
Physical Form Powder
Chemical Name or Material D-Fructose 1,6-bisphosphate trisodium salt octahydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.