Learn More
D(+)-Fenchone, 97%, ACROS Organics™
Brand: Acros Organics 366290250
Code : 29
Additional Details : CAS Number : 4695-62-9 Weight : 0.07500kg
Packaging | Glass bottle |
---|---|
Quantity | 25g |
Chemical Identifiers
4695-62-9 | |
152.24 | |
LHXDLQBQYFFVNW-XCBNKYQSSA-N | |
1201521 | |
(1R,4S)-2,2,4-trimethylbicyclo[2.2.1]heptan-3-one |
C10H16O | |
MFCD00070689 | |
+-fenchone, 1s-1,3,3-trimethylbicyclo 2.2.1 heptan-2-one, 1s-1,3,3-trimethyl-norbornan-2-one | |
CHEBI:165 | |
CC1(C2CCC(C2)(C1=O)C)C |
Specifications
D(+)-Fenchone | |
62°C | |
Glass bottle | |
0.95 | |
+ 59 | |
Colorless to Yellow | |
25g | |
4695-62-9 | |
C10H16O | |
07, IV, 212 | |
+-fenchone, 1s-1,3,3-trimethylbicyclo 2.2.1 heptan-2-one, 1s-1,3,3-trimethyl-norbornan-2-one | |
LHXDLQBQYFFVNW-XCBNKYQSSA-N | |
(1R,4S)-2,2,4-trimethylbicyclo[2.2.1]heptan-3-one | |
1201521 | |
152.24 | |
97% |
0.9500g/mL | |
Authentic | |
1.4610 to 1.464 | |
+59.00 (20.00°C neat) | |
63°C to 65°C (13.0mmHg) | |
5°C to 7°C | |
97% | |
96% min. (GC) | |
MFCD00070689 | |
15, 4000 | |
Solubility in water: practically insoluble in water. Other solubilities: soluble in ethanol,ether,common organic solvents | |
CC1(C2CCC(C2)(C1=O)C)C | |
152.24 | |
CHEBI:165 | |
Liquid |