missing translation for 'onlineSavingsMsg'
Learn More

D-Fenchol, TRC

Product Code. 30179310
Change view
Click to view available options
Quantity:
250 mg
5 g
Unit Size:
250mg
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30179310 250 mg 250mg
30179243 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30179310 Supplier Toronto Research Chemicals Supplier No. TRCF247455250MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2217-02-9
Molecular Formula C10 H18 O
Molecular Weight (g/mol) 154.25
Synonym (+)-Fenchyl alcohol, (1R,2R,4S)-1,3,3-Trimethylnorbornan-2-ol, Bicyclo[2.2.1]heptan-2-ol, 1,3,3-trimethyl-, (1R,2R,4S)-, Bicyclo[2.2.1]heptan-2-ol, 1,3,3-trimethyl-, (1R-endo)-, (1R,2R,4S)-1,3,3-Trimethylbicyclo[2.2.1]heptan-2-ol, (+)-Fenchol, (+)-α-Fenchol, (+)-α-Fenchyl alcohol, (1R)-endo-(+)-Fenchol, (1R)-endo-(+)-Fenchyl alcohol, (1R-endo)-Fenchol, D-Fenchol, D-Fenchyl alcohol, α-(+)-Fenchol, α-Fenchol, (+)-
IUPAC Name (1S,3R,4R)-2,2,4-trimethylbicyclo[2.2.1]heptan-3-ol
SMILES C[C@@]12CC[C@@H](C1)C(C)(C)[C@@H]2O

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C10H18O/c1-9(2)7-4-5-10(3,6-7)8(9)11/h7-8,11H,4-6H2,1-3H3/t7-,8-,10+/m0/s1
Formula Weight 154.1358
Chemical Name or Material (+)-Fenchyl Alcohol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.