missing translation for 'onlineSavingsMsg'
Learn More

D-Ethyl Gonendione, TRC

Product Code. 30380732
Change view
Click to view available options
Quantity:
100 mg
10 g
Unit Size:
100mg
10g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30380732 100 mg 100mg
30376270 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30380732 Supplier Toronto Research Chemicals Supplier No. TRCE918600100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 13-Ethylgon-4-ene-3,17-dione (Levodione; 18-Methyl-19-norandrost-4-ene-3,17-dione)
CAS 21800-83-9
Molecular Formula C19 H26 O2
Quantity 100 mg
Synonym 13-Ethylgon-4-ene-3,17-dione, Levonorgestrel Imp. L (Pharmeuropa), Levodione, 18-Methyl-19-norandrost-4-ene-3,17-dione
InChI Formula InChI=1S/C19H26O2/c1-2-19-10-9-15-14-6-4-13(20)11-12(14)3-5-16(15)17(19)7-8-18(19)21/h11,14-17H,2-10H2,1H3/t14-,15+,16+,17-,19-/m0/s1
SMILES CC[C@]12CC[C@H]3[C@@H](CCC4=CC(=O)CC[C@H]34)[C@@H]1CCC2=O
IUPAC Name (8R,9S,10R,13S,14S)-13-ethyl-1,2,6,7,8,9,10,11,12,14,15,16-dodecahydrocyclopenta[a]phenanthrene-3,17-dione
Molecular Weight (g/mol) 286.41
Formula Weight 286.1933
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.