missing translation for 'onlineSavingsMsg'
Learn More

D-Erythrose-13C4 (As a solution in water ~0.1M), TRC

Product Code. 30369649
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30369649 5 mg -
30384487 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30369649 Supplier Toronto Research Chemicals Supplier No. TRCE6501355MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula 13C4H8O4
Molecular Weight (g/mol) 124.08
Synonym (2R,3R)-2,3,4-TrihydroxyButanal-13C4
SMILES O[13C@H]1[13C@@H](O)[13CH](O)O[13CH2]1

Specifications

Solvent or Matrix Water
Quantity 5 mg
InChI Formula InChI=1S/C4H8O4/c5-2-1-8-4(7)3(2)6/h2-7H,1H2/t2-,3-,4?/m1/s1/i1+1,2+1,3+1,4+1
Concentration or Composition (by Analyte or Components) D-Erythrose-13C4 ~0.1M
Chemical Name or Material D-Erythrose-13C4
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.