missing translation for 'onlineSavingsMsg'
Learn More

D-erythro-Sphingosine (synthetic), TRC

Product Code. 30179563
Change view
Click to view available options
Quantity:
10 mg
100 mg
250 mg
Unit Size:
100mg
10mg
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30179563 10 mg 10mg
30179442 100 mg 100mg
30178950 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30179563 Supplier Toronto Research Chemicals Supplier No. TRCS68100010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-78-4
Molecular Formula C18 H37 N O2
Molecular Weight (g/mol) 299.49
Synonym D-Erythro-Sphingosine
IUPAC Name (E,2S,3R)-2-aminooctadec-4-ene-1,3-diol
SMILES CCCCCCCCCCCCC\C=C\[C@@H](O)[C@@H](N)CO

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C18H37NO2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-18(21)17(19)16-20/h14-15,17-18,20-21H,2-13,16,19H2,1H3/b15-14+/t17-,18+/m0/s1
Formula Weight 299.2824
Percent Purity >95%
Chemical Name or Material D-erythro-Sphingosine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.