missing translation for 'onlineSavingsMsg'
Learn More

D-erythro-Sphingosine, MedChemExpress

Product Code. 16452686
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16452686 25 mg 25mg
16433248 10 mg 10mg
16409796 50 mg 50mg
16404516 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16452686 Supplier Medchem Express Supplier No. HY101047

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress D-erythro-Sphingosine (Erythrosphingosine) is a very potent activator of p32-kinase with an EC50 of 8 μM, and inhibits protein kinase C (PKC). D-erythro-Sphingosine (Erythrosphingosine) is also a PP2A activator.

D-erythro-Sphingosine (Erythrosphingosine) is a very potent activator of p32-kinase with an EC50 of 8 μM, and inhibits protein kinase C (PKC). D-erythro-Sphingosine (Erythrosphingosine) is also a PP2A activator.

Specifications

Chemical Name or Material D-erythro-Sphingosine
Physical Form Solid
CAS 123-78-4
Color White
Quantity 25 mg
Purity Grade Notes Research
Molecular Formula C18H37NO2
For Use With (Application) Cancer-Kinase/protease
Synonym Erythrosphingosine erythro-C18-Sphingosine trans-4-Sphingenine
Solubility Information DMSO : 50 mg/mL (166.95 mM; ultrasonic and warming and heat to 60°C)
SMILES CCCCCCCCCCCCC/C=C/[C@@H](O)[C@@H](N)CO
Molecular Weight (g/mol) 299.49
Formula Weight 299.49
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.