missing translation for 'onlineSavingsMsg'
Learn More

D-erythro-Imidazoleglycerol Phosphate Monohydrate (~90%), TRC

Product Code. 30395980
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30395980 1 mg
30402354 10 mg
30408767 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30395980 Supplier Toronto Research Chemicals Supplier No. TRCI3500001MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 210241-69-3
Molecular Formula C6 H11 N2 O6 P . H2 O
Molecular Weight (g/mol) 256.15
IUPAC Name [(2R,3S)-2,3-dihydroxy-3-(4H-imidazol-4-yl)propyl] dihydrogen phosphate;hydrate
SMILES O.O[C@H](COP(=O)(O)O)[C@@H](O)C1C=NC=N1

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C6H11N2O6P.H2O/c9-5(2-14-15(11,12)13)6(10)4-1-7-3-8-4;/h1,3-6,9-10H,2H2,(H2,11,12,13);1H2/t4?,5-,6+;/m1./s1
Formula Weight 256.046
Chemical Name or Material D-erythro-Imidazoleglycerol Phosphate Monohydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.