Learn More
Invitrogen™ D-dimer Monoclonal Antibody (DD2)
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  Invitrogen™ MA120027
Product Code. 11509322
  • £455.00 / 100µg
Description
MA1-20027 detects D-Dimer in human samples.
Fibrinogen is the main protein of blood coagulation system. It consists of two identical subunits that contain three polypeptide chains: alpha, beta and gamma. The process of blood coagulation results in the activation of fibrinogen into fibrin by thrombin and fibrin polymerization. Fibrin clot is then digested by plasmin, and fibrin degradation products of different molecular weights are released into the bloodstream.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
D-dimer
Monoclonal
6.8 mg/mL
PBS with 0.09% sodium azide; pH 7.4
P02671
FGA
High molecular weight fibrin degradation products or synthetic peptides covering the cross-linked region of D-dimer gamma-chain.
100 μg
Primary
Human
Antibody
IgG2b
ELISA, Western Blot
DD2
Unconjugated
FGA
carboxy-terminal region; ENSMUSG00000059807; FGA; Fib; Fib2; fibrinogen A-alpha-chain; fibrinogen alpha chain; fibrinogen, A alpha polypeptide; Fibrinopeptide A; MGC119422; MGC119423; MGC119425
Mouse
Protein A
RUO
2243
4°C