missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ D-Dimer Human ELISA Kit

Sandwich ELISA Kit

Brand:  Invitrogen™ EHDDIMERX10

Additional Details : Weight : 2.50000kg

 View more versions of this product

Product Code. 17884190

  • £4280.00 / Pack of 10
Estimated Shipment: 26-06-2024
to see stock.
Add to basket

Description

Description

The Human D-Dimer ELISA quantitates Hu D-Dimer in human serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu D-Dimer. Principle of the method The Human D-Dimer solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

Fibrinogen is the main protein of blood coagulation system. It consists of two identical subunits that contain three polypeptide chains: alpha, beta and gamma. The process of blood coagulation results in the activation of fibrinogen into fibrin by thrombin and fibrin polymerization. Fibrin clot is then digested by plasmin, and fibrin degradation products of different molecular weights are released into the bloodstream.
Specifications

Specifications

P02671
0.08 pg/mL
Solid-phase sandwich ELISA
ELISA Kit
Plasma, Serum, Supernatant
10 x 96 assays
ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis
2243
<12%
Biotinylated Detection Antibody, Stop Solution, Pre-coated 96 well plate, SAV-HRP, Assay Diluent concentrate, Wash Buffer, Chromogen, Standard, Adhesive Plate Covers
10 x 96 Tests
Extracellular Matrix & Cell Adhesion
2°C to 8°C
Human
4 hr. 45 min.
0.082 to 60 pg/mL
0.08 pg/mL to 60 pg/mL
Biotin
Cell Structure & Organelle Proteins
This ELISA pair antibody detects human D-Dimer. Other species not determined.
Human
Colorimetric Microplate Reader
Fib2
<10%
HRP
RUO
Plasma, 1 μL; Serum, 1 μL; Supernatant, 100 μL
Fib2
1 hr. 20 min.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers