missing translation for 'onlineSavingsMsg'
Learn More

D-Dimer Antibody (DD5), Janelia Fluor™ 549, Novus Biologicals™

Mouse Monoclonal Antibody

£393.00

Specifications

Antigen D-Dimer
Clone DD5
Applications Western Blot
Classification Monoclonal
Conjugate Janelia Fluor 549
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18778182
View Documents
Novus Biologicals
NB120-10055JF549
0.2 mL
£393.00
0.20mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

D-Dimer Monoclonal antibody specifically detects D-Dimer in Human samples. It is validated for Western Blot, ELISA, ELISA
Specifications

Specifications

D-Dimer
Western Blot
Janelia Fluor 549
Mouse
Human
D Dimer
IgG2b
Protein A purified
DD5
Monoclonal
Purified
RUO
50mM Sodium Borate
Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with D-dimer.
Primary
Store at 4°C in the dark.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.