missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-Dimer Antibody (DD44), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
£393.00
Specifications
| Antigen | D-Dimer |
|---|---|
| Clone | DD44 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Conjugate | DyLight 594 |
Description
D-Dimer Monoclonal antibody specifically detects D-Dimer in Human samples. It is validated for Western Blot, ELISASpecifications
| D-Dimer | |
| Western Blot | |
| DyLight 594 | |
| Mouse | |
| Human | |
| D Dimer | |
| IgG2b | |
| Protein A purified |
| DD44 | |
| Monoclonal | |
| Purified | |
| RUO | |
| 50mM Sodium Borate | |
| Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with mixture of D-dimer and high molecular weight fibrin degradation products. | |
| Primary | |
| Store at 4°C in the dark. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title