missing translation for 'onlineSavingsMsg'
Learn More

D-(+)-Dextrose, Anhydrous

Product Code. 15825128 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
5 lb.
25 lb.
Packaging:
Plastic bottle
Unit Size:
11339.8g
2268g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15825128 25 lb. Plastic bottle 11339.8g
15815128 5 lb. Plastic bottle 2268g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15825128 Supplier Thermo Scientific Supplier No. J14535.Q1
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 50-99-7 | C6H12O6 | 180.156 g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-99-7
Molecular Formula C6H12O6
Molecular Weight (g/mol) 180.156
MDL Number MFCD00063774
InChI Key GZCGUPFRVQAUEE-SLPGGIOYSA-N
Synonym 2 r,3 s,4 r,5 r-2,3,4,5,6-pentahydroxyhexanal, d +-glucose, aldehydo-d-glucose, glucose, anhydrous, dextrose, anhydrous, d-glucose, anhydrous, glucosteril, anhydrous dextrose, dextrose solution, d-glucose in linear form
PubChem CID 107526
ChEBI CHEBI:42758
IUPAC Name (2 R,3 S,4 R,5 R)-2,3,4,5,6-pentahydroxyhexanal
SMILES C(C(C(C(C(C=O)O)O)O)O)O

Specifications

Melting Point 147°C to 153°C
Packaging Plastic bottle
Linear Formula CH2OH(CHOH)4CHO
Quantity 25 lb.
Beilstein 1281608
Formula Weight 180.16
Physical Form Crystalline Powder
Chemical Name or Material D-(+)-Dextrose

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.