missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IBA Lifesciences 10x Buffer E - Elution Buffer
Shop All IBA Lifesciences Products
Click to view available options
Quantity:
25 mL
Unit Size:
Each
Description
- 10x Buffer E is a 10x concentrated Strep-Tactin™ elution buffer.
- The contained desthiobiotin is a modified form of biotin that displaces Strep-tag™II and Twin-Strep-tag™ fusion proteins from the modified biotin-binding pocket of Strep-Tactin™ in a competitive manner, resulting in a mild elution.
- Please note that 10x Buffer E is not applicable for elution from Strep-Tactin™XT resins.
Specifications
Specifications
| Chemical Name or Material | Desthiobiotin Buffer E |
| Description | 10 X, Buffer E, Contains Tris pH 8, NaCl and EDTA |
| Concentration | 10 X |
| For Use With (Application) | Ready-to-Use for Elution of Strep-Tag II Proteins From The Biotin-Binding Site of Strep-Tactin Resin |
| Quantity | 25 mL |
| Formulation | 10 X, Buffer E, Contains Tris pH 8, NaCl and EDTA |
Safety and Handling
Product Identifier
- Buffer E (10x), Strep-Tactin Elution Buffer with Desthiobiotin
For Research Use Only.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction