Learn More
IBA Lifesciences Desthiobiotin
Desthiobiotin powder for preparation of Strep-Tactin™ elution buffer
Brand:  IBA Lifesciences 2-1000-005
Code : LE
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 12763064
  • £1350.00 / Each
Estimated Shipment: 14-05-2024
to see stock.
Description
Description
Desthiobiotin is a modified form of biotin that displaces Strep-tag™II and Twin-Strep-tag™ fusion proteins from the modified biotin-binding pocket of Strep-Tactin™ in a competitive manner, resulting in a mild elution.
Desthiobiotin powder can be used for preparation of Strep-Tactin™ elution buffer, Buffer E.
Please note that desthiobiotin is not applicable for elution from Strep-Tactin™XT resins.
Specifications
Specifications
5 g
For Elution of Strep-Tag II Proteins From Strep-Tactin
Desthiobiotin