missing translation for 'onlineSavingsMsg'
Learn More

D-Cystine, 98%

Product Code. 10627682 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10627682 250 mg Glass Bottle 250mg
10235090 1 g Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10627682 Supplier Thermo Scientific Acros Supplier No. 133982500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 349-46-2 | C6H12N2O4S2 | 240.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 349-46-2
Molecular Formula C6H12N2O4S2
Molecular Weight (g/mol) 240.29
MDL Number MFCD00002610
InChI Key LEVWYRKDKASIDU-IMJSIDKUSA-N
Synonym d-cystine, 2-amino-3-2s-2-amino-2-carboxyethyl disulfanyl propanoic acid
PubChem CID 6857538
ChEBI CHEBI:35494
IUPAC Name (2S)-2-amino-3-[[(2S)-2-amino-2-carboxyethyl]disulfanyl]propanoic acid
SMILES N[C@@H](CSSC[C@H](N)C(O)=O)C(O)=O

Specifications

Melting Point 260.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass Bottle
Linear Formula [-SCH2CH(NH2)CO2H]2
Specific Rotation + 214.00
Merck Index 15, 2779
Sulfated Ash 0.1% max.
Solubility Information Solubility in water: 0.057g/L (25°C).
Quantity 250 mg
Formula Weight 240.29
Percent Purity 98%
Physical Form Fine Crystalline Powder
Chemical Name or Material D-Cystine, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.