missing translation for 'onlineSavingsMsg'
Learn More

D-Cysteine, TRC

Product Code. 30179668
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30179668 1 g 1g
30179378 25 g 25g
30179394 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30179668 Supplier Toronto Research Chemicals Supplier No. TRCC9950201G
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 921-01-7
Molecular Formula C3 H7 N O2 S
Molecular Weight (g/mol) 121.16
Synonym D-Cysteine (9CI, ACI), Cysteine, D- (8CI), (2S)-2-Amino-3-sulfanylpropanoic acid, (S)-2-Amino-3-mercaptopropanoic acid, (S)-Cysteine
IUPAC Name (2S)-2-amino-3-sulfanylpropanoic acid
SMILES N[C@H](CS)C(=O)O

Specifications

InChI Formula InChI=1S/C3H7NO2S/c4-2(1-7)3(5)6/h2,7H,1,4H2,(H,5,6)/t2-/m1/s1
Quantity 1 g
Formula Weight 121.0197
Shipping Condition Room Temperature
Chemical Name or Material D-Cysteine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.