Learn More
D-Cysteine, 98%, Thermo Scientific Chemicals
£50.80 - £219.00
Chemical Identifiers
CAS | 921-01-7 |
---|---|
Molecular Formula | C3H7NO2S |
Molecular Weight (g/mol) | 121.15 |
MDL Number | MFCD00066461 |
InChI Key | XUJNEKJLAYXESH-RUOZJJLTNA-N |
Synonym | d-cysteine, d-cystein, 2s-2-amino-3-sulfanylpropanoic acid, d-zystein, cysteine, d, s-2-amino-3-mercaptopropanoic acid, d-amino-3-mercaptopropionic acid, d-cys, h-d-cys-oh, cysteine |
PubChem CID | 92851 |
ChEBI | CHEBI:16375 |
IUPAC Name | (2S)-2-amino-3-sulfanylpropanoic acid |
SMILES | N[C@H](CS)C(O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15439138
|
Thermo Scientific Alfa Aesar
H62627.03 |
1 g |
£50.80
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15449138
|
Thermo Scientific Alfa Aesar
H62627.06 |
5 g |
£219.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
D-Cysteine is an important source of sulfide in human metabolism. It is the precursor for glutathione. It plays important role in protein structure and in metal binding.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsD-Cysteine is an important source of sulfide in human metabolism. It is the precursor for glutathione. It plays important role in protein structure and in metal binding.
Solubility
Soluble in water (partly).
Notes
Air sensitive. Store under inert gas. Store away from oxidizing agents, air.
Chemical Identifiers
921-01-7 | |
121.15 | |
XUJNEKJLAYXESH-RUOZJJLTNA-N | |
92851 | |
(2S)-2-amino-3-sulfanylpropanoic acid |
C3H7NO2S | |
MFCD00066461 | |
d-cysteine, d-cystein, 2s-2-amino-3-sulfanylpropanoic acid, d-zystein, cysteine, d, s-2-amino-3-mercaptopropanoic acid, d-amino-3-mercaptopropionic acid, d-cys, h-d-cys-oh, cysteine | |
CHEBI:16375 | |
N[C@H](CS)C(O)=O |
Specifications
230°C (decomposition) | |
C3H7NO2S | |
1721407 | |
Soluble in water (partly). | |
N[C@H](CS)C(O)=O | |
121.15 | |
CHEBI:16375 | |
98% | |
Solid |
921-01-7 | |
MFCD00066461 | |
d-cysteine, d-cystein, 2s-2-amino-3-sulfanylpropanoic acid, d-zystein, cysteine, d, s-2-amino-3-mercaptopropanoic acid, d-amino-3-mercaptopropionic acid, d-cys, h-d-cys-oh, cysteine | |
XUJNEKJLAYXESH-RUOZJJLTNA-N | |
(2S)-2-amino-3-sulfanylpropanoic acid | |
92851 | |
121.16 | |
Air Sensitive | |
D-Cysteine |
Safety and Handling
GHS H Statement
H302
Harmful if swallowed.
P501c
Warning
missing translation for 'einecsNumber' : 213-062-0
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only