missing translation for 'onlineSavingsMsg'
Learn More

D-Cycloserine, TRC

Product Code. 30134486
Change view
Click to view available options
Quantity:
100 mg
250 mg
5 g
Unit Size:
100mg
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134486 5 g 5g
30010408 250 mg 250mg
30005654 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134486 Supplier Toronto Research Chemicals Supplier No. TRCC9888155G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68-41-7
Molecular Formula C3 H6 N2 O2
Molecular Weight (g/mol) 102.09
Synonym Cycloserine, D-(+)-Cycloserine, 3-Isoxazolidinone, 4-amino-, (4R)-, 3-Isoxazolidinone, 4-amino-, (+)- (8CI), 3-Isoxazolidinone, 4-amino-, (R)-, (+)-Cycloserine, (4R)-4-Amino-3-isoxazolidinone, (R)-(+)-Cycloserine, (R)-4-Amino-3-isoxazolidinone, (R)-Cycloserine, Closerin, Closina, Cyclo-D-serine, Cyclorin, Cycloserine, D-4-Amino-3-isoxazolidinone, D-4-Amino-3-isoxazolidone, D-CS, D-Cycloserine, Farmiserina, Micoserina, Miroseryn, NSC 154851, NSC 76029, Novoserin, Orientomycin, Oxamycin, PA 94, Seromycin, Tisomycin, Wasserina, d-4-Amino-3-isoxazolidinone, α-Cycloserine
IUPAC Name (4R)-4-amino-1,2-oxazolidin-3-one
SMILES N[C@@H]1CONC1=O

Specifications

InChI Formula InChI=1S/C3H6N2O2/c4-2-1-7-5-3(2)6/h2H,1,4H2,(H,5,6)/t2-/m1/s1
Quantity 5 g
Formula Weight 102.0429
Shipping Condition Room Temperature
Chemical Name or Material Cycloserine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.