missing translation for 'onlineSavingsMsg'
Learn More

D-Cycloserine, MedChemExpress

Product Code. 16488066
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16488066 100 mg 100mg
16471996 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16488066 Supplier Medchem Express Supplier No. HYB0030
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress D-Cycloserine is an antibiotic which targets sequential bacterial cell wall peptidoglycan biosynthesis enzymes. D-Cycloserine is a partial NMDA agonist that can improve cognitive functions. D-Cycloserine can be used for multidrug-resistant tuberculosis research.

D-Cycloserine is an antibiotic which targets sequential bacterial cell wall peptidoglycan biosynthesis enzymes. D-Cycloserine is a partial NMDA agonist that can improve cognitive functions. D-Cycloserine can be used for multidrug-resistant tuberculosis research.

Specifications

Chemical Name or Material D-Cycloserine
Physical Form Powder
CAS 68-41-7
Color Off-White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C3H6N2O2
For Use With (Application) COVID-19-immunoregulation
Solubility Information H2O : 25 mg/mL (244.88 mM; Need ultrasonic) ∣DMSO : 1 mg/mL (9.80 mM; Need warming)
SMILES O=C1[C@H](N)CON1
Molecular Weight (g/mol) 102.09
Formula Weight 102.09
Percent Purity 98.28%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.