missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D-Cycloserine, 98%

Product Code. 10683062 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10683062 1 g Glass bottle 1g
10450501 5 g Glass bottle 5g
10646734 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10683062 Supplier Thermo Scientific Chemicals Supplier No. 228480010
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

D-Cycloserine, CAS # 68-41-7, is a cyclic amino acid analog of D-alanine, originally isolated from Streptomyces species. It is synthetically formulated and exhibits antibiotic properties through its inhibition of bacterial cell wall synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General description

  • D-Cycloserine was originally isolated from Streptomyces orchidaceus and is currently formulated as an oral antibiotic
  • It is incompatible with strong oxidizing agents but stable under basic conditions, with the greatest stability at pH = 11.5
  • D-Cycloserine inhibits L-alanine racemase and D-alanine:D-alanine ligase, 2 enzymes responsible for bacterial cell wall synthesis
  • When ingested, D-Cycloserine traverses the blood-brain barrier, potentially causing neurological side effects such as, dizziness, nausea, depression, tremors, etc.

Applications

  • D-Cycloserine is a broad-spectrum antibiotic used as a second line agent for treatment of drug-resistant tuberculosis, always in combination with other antituberculosis agents
  • It is an excitatory amino acid and partial agonist of the NMDA receptor in the central nervous system (CNS), thus playing a potential role in the amelioration of neuropathic pain
  • D-Cycloserine has also been shown to play a role in neural connectivity, and is therefore used to study treatment applications in anxiety disorders and schizophrenia
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68-41-7
Molecular Formula C3H6N2O2
Molecular Weight (g/mol) 102.09
MDL Number MFCD00005353
InChI Key DYDCUQKUCUHJBH-UWTATZPHSA-N
Synonym d-cycloserine, cycloserine, seromycin, orientomycin, oxamycin, cyclorin, cyclo-d-serine, cicloserina, farmiserina, miroseryn
PubChem CID 6234
ChEBI CHEBI:40009
IUPAC Name (4R)-4-amino-1,2-oxazolidin-3-one
SMILES C1C(C(=O)NO1)N

Specifications

Absorbance Condensation products 0.8 max. (c=0.04, 0.1N NaOH) at 285nm
Melting Point 147.0°C
Color White to Yellow
pH 5.5 to 6.5 (10% soln.)
Ignition Residue 0.5% max. ()
Loss on Drying 1% max. (60°C, 3 hrs) (vacuum)
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 1 g
Merck Index 15,2749
Specific Rotation + 111.00
Solubility Information Solubility in water: soluble. Other solubilities: slightly soluble in methanol and propylene glycol
Formula Weight 102.09
Percent Purity 98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material D-Cycloserine, 98+%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.