missing translation for 'onlineSavingsMsg'
Learn More

Melford D-Cycloserine

Product Code. 30018207
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30018207 5 g 5g
30018206 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30018207 Supplier Melford Supplier No. C810005.0
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

D-cycloserine is an antibiotic and structural analog of the amino acid D-alanine.

D-cycloserine is an antibiotic and structural analog of the amino acid D-alanine. It inhibits cell wall synthesis in certain bacteria, including Mycobacterium tuberculosis. While it was initially identified as an antibiotic, D-cycloserine has gained attention for its activity as a partial agonist at the N-methyl-D-aspartate (NMDA) receptor, a type of glutamate receptor in the brain. Researchers study the mechanisms of neurotransmission and synaptic plasticity due to its interactions with NMDA receptors.

Specifications

Chemical Name or Material D-Cycloserine
CAS 68-41-7
Quantity 5 g
Molecular Formula C3H6N2O2
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.